Ethyl 2-hydroxyethyl sulfide, 98%
Ethyl 2-hydroxyethyl sulfide, 98%
Ethyl 2-hydroxyethyl sulfide, 98%
Thermo Scientific Chemicals

Ethyl 2-hydroxyethyl sulfide, 98%

CAS: 110-77-0 | C4H10OS | 106.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
118551000100 gGlass bottle
Catalog number 118551000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-77-0
IUPAC Name2-(ethylsulfanyl)ethan-1-ol
Molecular FormulaC4H10OS
InChI KeyLNRIEBFNWGMXKP-UHFFFAOYSA-N
SMILESCCSCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4859 to 1.4879 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only