Ethyl indole-2-carboxylate, 97%
Ethyl indole-2-carboxylate, 97%
Ethyl indole-2-carboxylate, 97%
Acros Organics

Ethyl indole-2-carboxylate, 97%

CAS: 3770-50-1 | C11H11NO2 | 189.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 118600250
also known as 11860
HSN 29339900
Price (INR)
36,929.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
36,929.00
Each
Chemical Identifiers
CAS3770-50-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
GC>=96.0 %
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
Melting point119°C to 125°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only