Ethyl 2-mercaptoacetate, 99%
Ethyl 2-mercaptoacetate, 99%
Ethyl 2-mercaptoacetate, 99%
Thermo Scientific Chemicals

Ethyl 2-mercaptoacetate, 99%

CAS: 623-51-8 | C4H8O2S | 120.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 118671000
Price (EUR)
63,50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
63,50
Each
Chemical Identifiers
CAS623-51-8
IUPAC Nameethyl 2-sulfanylacetate
Molecular FormulaC4H8O2S
InChI KeyPVBRSNZAOAJRKO-UHFFFAOYSA-N
SMILESCCOC(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4560 to 1.4590 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only