RPMI 1640 Medium
Gibco™

RPMI 1640 Medium

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park MemorialRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
118751356 x 1000 mL
11875101100 mL
1187512720 x 100 mL
11875093500 mL
1187511910 x 500 mL
118750851000 mL
Catalog number 11875135
Price (TWD)
7,210.00
Online offer
Ends: 31-Dec-2026
10,300.00
Save 3,090.00 (30%)
Each
Quantity:
6 x 1000 mL
Customize this product
Price (TWD)
7,210.00
Online offer
Ends: 31-Dec-2026
10,300.00
Save 3,090.00 (30%)
Each

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red


The complete formulation is available.

Using RPMI
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity6 x 1000 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (7)

Citations & References
Abstract
Smac agonists sensitize for Apo2L/TRAIL- or anticancer drug-induced apoptosis and induce regression of malignant glioma in vivo.
Authors: Fulda Simone; Wick Wolfgang; Weller Michael; Debatin Klaus-Michael;
Journal:Nat Med
PubMed ID:12118245
'A major concern in cancer therapy is resistance of tumors such as glioblastoma to current treatment protocols. Here, we report that transfer of the gene encoding second mitochondria-derived activator of caspase (Smac) or Smac peptides sensitized various tumor cells in vitro and malignant glioma cells in vivo for apoptosis induced ... More
Size, concentration and incubation time dependence of gold nanoparticle uptake into pancreas cancer cells and its future application to X-Ray Drug Delivery System.
Authors:Trono JD, Mizuno K, Yusa N, Matsukawa T, Yokoyama K, Uesaka M
Journal:J Radiat Res
PubMed ID:21187668
'One of the restrictions in the potential use of gold markers for medical imaging/tracking of harder tumors is its size. We propose to use gold nanoparticles which, due to its small size, can be administered conveniently via intravenous injection. One of the factors that determine the clinical utility of nanoparticles ... More
Cell docking and on-chip monitoring of cellular reactions with a controlled concentration gradient on a microfluidic device.
Authors: Yang Mengsu; Li Cheuk-Wing; Yang Jun;
Journal:Anal Chem
PubMed ID:12199565
We have developed a microfluidic device for on-chip monitoring of cellular reactions. The device consists of two primary analytical functions: control of cell transport and immobilization, and dilution of an analyte solution to generate a concentration gradient. In this device, a dam structure in parallel to the fluid flow was ... More
EMR4, a Novel Epidermal Growth Factor (EGF)-TM7 Molecule Up-regulated in Activated Mouse Macrophages, Binds to a Putative Cellular Ligand on B Lymphoma Cell Line A20.
Authors: Stacey Martin; Chang Gin-Wen; Sanos Stephanie L; Chittenden Laura R; Stubbs Lisa; Gordon Siamon; Lin Hsi-Hsien;
Journal:J Biol Chem
PubMed ID:12023293
A novel member of the EGF-TM7 family, mEMR4, was identified and characterized. The full-length mouse EMR4 cDNA encodes a predicted 689-amino acid protein containing two epidermal growth factor (EGF)-like modules, a mucin-like spacer domain, and a seven-transmembrane domain with a cytoplasmic tail. Genetic mapping established that mEMR4 is localized in ... More
Malignant effusions and immunogenic tumour-derived exosomes.
Authors: Andre Fabrice; Schartz Noel E C; Movassagh Mojgan; Flament Caroline; Pautier Patricia; Morice Philippe; Pomel Christophe; Lhomme Catherine; Escudier Bernard; Le Chevalier Thierry; Tursz Thomas; Amigorena Sebastian; Raposo Graca; Angevin Eric; Zitvogel Laurence;
Journal:Lancet
PubMed ID:12147364
BACKGROUND: Exosomes derived from tumours are small vesicles released in vitro by tumour cell lines in culture supernatants. To assess the role of these exosomes in vivo, we examined malignant effusions for their presence. We also investigated whether these exosomes could induce production of tumour-specific T cells when pulsed with ... More