Ethyl oxamate, 99%
Ethyl oxamate, 99%
Ethyl oxamate, 99%
Thermo Scientific Chemicals

Ethyl oxamate, 99%

CAS: 617-36-7 | C4H7NO3 | 117.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
Catalog number 118790250
Price (EUR)
23,10
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
23,10
Each
Chemical Identifiers
CAS617-36-7
IUPAC Nameethyl carbamoylformate
Molecular FormulaC4H7NO3
InChI KeyRZMZBHSKPLVQCP-UHFFFAOYSA-N
SMILESCCOC(=O)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Assay>=98.5 %
Appearance (Form)Crystalline powder, crystals and/or chunks
Melting point114°C to 119°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only