DMEM, low glucose, pyruvate, 500 mL - FAQs
DMEM, low glucose, pyruvate, 500 mL - FAQs
View additional product information for DMEM, low glucose, pyruvate - FAQs (11885076, 11885092, 11885084)
3 product FAQs found
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf life of the DMEM, high glucose, pyruvate medium once the bottle is opened and the medium is supplemented?
We do not provide stability data for the product once it is opened as it would depend on the usage and storage conditions.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is it necessary to store DMEM, high glucose, pyruvate medium in the dark?
Yes, the medium should be stored in the dark because there are some components in the medium such as HEPES, Tryptophan, and Riboflavin, etc. that are sensitive to light.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.