Ethyl N-piperazinecarboxylate, 99%
Ethyl N-piperazinecarboxylate, 99%
Ethyl N-piperazinecarboxylate, 99%
Thermo Scientific Chemicals

Ethyl N-piperazinecarboxylate, 99%

CAS: 120-43-4 | C7H14N2O2 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 118871000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS120-43-4
IUPAC Nameethyl piperazine-1-carboxylate
Molecular FormulaC7H14N2O2
InChI KeyLNOQURRKNJKKBU-UHFFFAOYSA-N
SMILESCCOC(=O)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Refractive index1.4760 to 1.4790 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only