Ethyl pyruvate, 98%
Ethyl pyruvate, 98%
Ethyl pyruvate, 98%
Thermo Scientific Chemicals

Ethyl pyruvate, 98%

CAS: 617-35-6 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 118945000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS617-35-6
IUPAC Nameethyl 2-oxopropanoate
Molecular FormulaC5H8O3
InChI KeyXXRCUYVCPSWGCC-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only