4-Ethylresorcinol, 98%
4-Ethylresorcinol, 98%
4-Ethylresorcinol, 98%
Thermo Scientific Chemicals

4-Ethylresorcinol, 98%

CAS: 2896-60-8 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 118950050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2896-60-8
IUPAC Name4-ethylbenzene-1,3-diol
Molecular FormulaC8H10O2
InChI KeyVGMJYYDKPUPTID-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point95°C to 99°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only