SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal
Invitrogen™

SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoN.º de reaccionesIncluye
119001132000 ReactionsSuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
1190000150 ReactionsSuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
11900002500 ReactionsSuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
119000032000 ReactionsSuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
1190011150 ReactionsSuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
11900112500 ReactionsSuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
119000041250 ReactionsSuperScript V VILO Master Mix (Primer-free) only
Número de catálogo 11900113
Precio (USD)
23.644,41
Each
N.º de reacciones:
2000 Reactions
Incluye:
SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
Precio (USD)
23.644,41
Each

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy. Powered by SuperScript™ V Reverse Transcriptase, the optimized master mix combines robust performance across a broad range of RNA samples with simplified reaction setup, enhanced reagent stability, and greater workflow flexibility. SuperScript V VILO Master Mix, Primer-Free is also available with ezDNase™ Master Mix for fast and convenient genomic DNA removal prior to reverse transcription.

Features of SuperScript V VILO Master Mix, Primer-Free include:

  • Broad RNA input range—supports sensitive cDNA synthesis from 0.1 pg to 5 μg of total RNA
  • Robust performance with challenging RNA—efficient cDNA synthesis from highly structured, GC-rich, degraded and inhibitor-containing RNA
  • Fast cDNA synthesis—complete reverse transcription in as little as 1-10 minutes
  • Enhanced reagent stability—supports ambient shipping and room-temperature storage for up to 1 month
  • Simple, flexible workflow—ready-to-use format simplifies reaction setup, while room-temperature assembly and extended stability of assembled reactions enable greater workflow flexibility

SuperScript V VILO Master Mix, Primer-Free combines SuperScript V RT, RNase inhibitor, buffer, and dNTPs in a single formulation while allowing users to select and optimize primers for their specific application. This flexibility supports gene-specific primers, oligo(dT), random primers, or customized primer combinations while maintaining the convenience of a ready-to-use master mix.

SuperScript V VILO Master Mix, Primer-Free with gDNA Removal allows streamlined genomic DNA removal step. The dsDNA-specific ezDNase enzyme removes contaminating genomic DNA in as little as 2 minutes without affecting RNA.

SuperScript V VILO Master Mix, Primer-Free also features a DTT-free, animal origin–free formulation for improved sustainability and simplified reagent handling.

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Tipo de producto finalFirst-Strand cDNA
FormatoMaster Mix
IncluyeSuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
Inhibitor Tolerance (Simple)High
N.º de reacciones2000 Reactions
Temperatura óptima de reacción50°C
Tipo de productoReverse Transcription Reagent
Tipo de reactivoReverse Transcription
Transcriptasa inversaSuperScript V
Actividad de la ribonucleasa HReduced
Sensibilidad0.1 pg total RNA
Condiciones de envíoDry Ice, Wet Ice, Ambient
Tamaño (producto final)Up to 20 kb
Material de partidaRNA
TécnicaReverse Transcription
TermoestabilidadUp to 65°
Concentración5X
Método de detecciónqPCR, NGS, PCR
Para utilizar con (aplicación)Gene Expression, Real Time PCR (qPCR), RT-PCR, cDNA Synthesis, Next-Generation Sequencing (NGS)
FormularioLiquid
GC-Rich PCR PerformanceHigh
Etiqueta o tinteNot Applicable
Método de PCR2-step RT-qPCR
Velocidad de reacción10 min.
Unit SizeEach
Contenido y almacenamiento
• Four 500 reaction packages of SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal (Cat. No. 11900112)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 5 months
• Room temperature for up to 1 month

Preguntas frecuentes

What are the main differences between Invitrogen SuperScript V Reverse Transcriptase and Invitrogen SuperScript V VILO Master Mix?

Invitrogen SuperScript V Reverse Transcriptase is supplied as a stand-alone enzyme with reaction buffer and supports flexible reaction optimization. Invitrogen SuperScript V VILO Master Mix combines all required reaction components into a single formulation to simplify workflow setup and reduce pipetting steps.

Can Invitrogen SuperScript V Reverse Transcriptase products support automated workflows?

Yes. Invitrogen SuperScript V Reverse Transcriptase products include features such as room-temperature reaction setup, benchtop stability of assembled reactions, and simplified workflow options that can support automated and high-throughput workflows.

Can Invitrogen SuperScript V Reverse Transcriptase products support low-input RNA workflows?

Yes. Invitrogen SuperScript V Reverse Transcriptase products are designed to support reverse transcription from low-input RNA samples, including inputs as low as 0.1 pg of total RNA.

Which Invitrogen SuperScript V Reverse Transcriptase product should I choose for RT-qPCR workflows?

For two-step RT-qPCR workflows, Invitrogen SuperScript V VILO Master Mix reagent (Cat. No. 11800001) provides an all-in-one formulation that simplifies reaction setup. Users who prefer custom primer selection may choose Invitrogen SuperScript V VILO Master Mix, Primer-Free (Cat. No. 11900001) or Invitrogen SuperScript V Reverse Transcriptase (Cat. No. 19090050).

Can reaction volumes be scaled when using Invitrogen SuperScript V Reverse Transcriptase products?

Yes. Reaction volumes may be scaled up to 100 µL when using Invitrogen SuperScript V Reverse Transcriptase products provided that reagent ratios are maintained and workflow performance is verified.