Farnesol, 96%, mixture of isomers
Farnesol, 96%, mixture of isomers
Farnesol, 96%, mixture of isomers
Thermo Scientific Chemicals

Farnesol, 96%, mixture of isomers

CAS: 4602-84-0 | C15H26O | 222.37 g/mol
数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 119121000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4602-84-0
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Density(20/4°C) 0.884 to 0.89 kg/l
Infrared spectrumConforms
GC>=95 % (sum of geometric isomers)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only