Iron(III) acetylacetonate, 99+%
Iron(III) acetylacetonate, 99+%
Iron(III) acetylacetonate, 99+%
Thermo Scientific Chemicals

Iron(III) acetylacetonate, 99+%

CAS: 14024-18-1 | C15H21FeO6 | 353.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 119130010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14024-18-1
IUPAC Nameiron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21FeO6
InChI KeyAQBLLJNPHDIAPN-LNTINUHCSA-K
SMILES[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration Iodometric>=99.0 %
Appearance (Color)Dark orange to dark red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only