Fisetin
Thermo Scientific Chemicals

Fisetin

CAS: 528-48-3 | C15H10O6 | 286.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 119150050
Price (USD)
588.65
Online Exclusive
654.00
Save 65.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
588.65
Online Exclusive
654.00
Save 65.35 (10%)
Each
Chemical Identifiers
CAS528-48-3
IUPAC Name2-(3,4-dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one
Molecular FormulaC15H10O6
InChI KeyXHEFDIBZLJXQHF-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<6 %
Appearance (Color)White to amber to dark green
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=90.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only