Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=97.5 %
Appearance (Color)Yellow to yellow-green to beige to light grey
Infrared spectrumConforms
Melting point106°C to 111°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.