Fluoranthene, 98%
Fluoranthene, 98%
Fluoranthene, 98%
Acros Organics

Fluoranthene, 98%

CAS: 206-44-0 | C16H10 | 202.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 119171000
also known as 11917
HSN 29029090
Price (INR)
20,560.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
20,560.00
Each
Chemical Identifiers
CAS206-44-0
IUPAC Namefluoranthene
Molecular FormulaC16H10
InChI KeyGVEPBJHOBDJJJI-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=97.5 %
Appearance (Color)Yellow to yellow-green to beige to light grey
Infrared spectrumConforms
Melting point106°C to 111°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only