2-Fluoroaniline, 99+%
2-Fluoroaniline, 99+%
2-Fluoroaniline, 99+%
Thermo Scientific Chemicals

2-Fluoroaniline, 99+%

CAS: 348-54-9 | C6H6FN | 111.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 119271000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS348-54-9
IUPAC Name2-fluoroaniline
Molecular FormulaC6H6FN
InChI KeyFTZQXOJYPFINKJ-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to brown
AppearanceMay darken on storage
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only