DRI™ Gravity-Detect™ Specimen Validity Test
DRI™ Gravity-Detect™ Specimen Validity Test
Thermo Scientific™
DRI™ Gravity-Detect™ Specimen Validity Test
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test.
Have Questions?
Catalog NumberQuantity
11942 x 500 mL
Catalog number 1194
Price (USD)
-
Request A Quote
Quantity:
2 x 500 mL
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.
The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • Associated calibrators and controls offered in convenient sizes
  • Complete and automated way to test for sample integrity and adulteration
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDRI Gravity-Detect Assay Specimen Validity Test
Quantity2 x 500 mL
Sample TypeUrine
Shelf LifeUntil Label Expiry Date
TypeReagent
Detectable AnalytesSpecific Gravity
Unit SizeEach
Contents & Storage
Room Temperature