3-Fluorobenzyl bromide, 95%
3-Fluorobenzyl bromide, 95%
3-Fluorobenzyl bromide, 95%
Thermo Scientific Chemicals

3-Fluorobenzyl bromide, 95%

CAS: 456-41-7 | C7H6BrF | 189.027 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 119400050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS456-41-7
IUPAC Name1-(bromomethyl)-3-fluorobenzene
Molecular FormulaC7H6BrF
InChI KeySCBZBMXPJYMXRC-UHFFFAOYSA-N
SMILESFC1=CC=CC(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %
Refractive index1.5450 to 1.5480 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only