Interested in a bulk or custom version of this product? Request pricing.

SLAMF6 Recombinant Human Protein, His Tag

Catalog number: 11945H08H25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SLAM family member 6 (SLAMF6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human SLAMF6 (Q96DU3-1) (Met 1-Met 226) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind biotinylated recombinant human SH2D1A in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP11-528G1.1, FLJ50657, KALI, KALIb, Ly108, MGC104953, NTB-A, NTBA, SF2000
Gene Symbol: SLAMF6
Molecular Weight: 24.3 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Name: SLAM family member 6
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Research Category: Immunology, Signal Transduction
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products