1-Fluoro-4-nitrobenzene, 99%
1-Fluoro-4-nitrobenzene, 99%
1-Fluoro-4-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Fluoro-4-nitrobenzene, 99%

CAS: 350-46-9 | C6H4FNO2 | 141.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 119491000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS350-46-9
IUPAC Name1-fluoro-4-nitrobenzene
Molecular FormulaC6H4FNO2
InChI KeyWFQDTOYDVUWQMS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Refractive index1.5302 to 1.5322 (20°C, 589 nm)
Appearance (Color)Clear yellow to dark brown
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only