2-Fluorophenol, 98%
2-Fluorophenol, 98%
2-Fluorophenol, 98%
Thermo Scientific Chemicals

2-Fluorophenol, 98%

CAS: 367-12-4 | C6H5FO | 112.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 119541000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS367-12-4
IUPAC Name2-fluorophenol
Molecular FormulaC6H5FO
InChI KeyHFHFGHLXUCOHLN-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Refractive index1.5134 to 1.5154 (20°C, 589 nm)
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only