2-Fluoropyridine, 98%
2-Fluoropyridine, 98%
2-Fluoropyridine, 98%
Thermo Scientific Chemicals

2-Fluoropyridine, 98%

CAS: 372-48-5 | C5H4FN | 97.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
50 mL
Packaging:
Glass bottle
Catalog number 119630050
Price (USD)
26.65
Online Exclusive
29.70
Save 3.05 (10%)
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.70
Save 3.05 (10%)
Each
Chemical Identifiers
CAS372-48-5
IUPAC Name2-fluoropyridine
Molecular FormulaC5H4FN
InChI KeyMTAODLNXWYIKSO-UHFFFAOYSA-N
SMILESFC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4670 to 1.4690 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only