Platinum™ Taq Green Hot Start DNA Polymerase

Catalog number: 11966026

Invitrogen™  Related applications: Genotyping & Genomic Profiling | Methylation Analysis | PCR | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Invitrogen™ Platinum™ Taq Green Hot Start DNA Polymerase provides Platinum Taq DNA Polymerase with a 10X Green PCR Buffer. Platinum Taq DNA Polymerase is a convenient and reliable "hot start" thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The 10X Green PCR Buffer contains a density reagent and two tracking dyes and offers the convenience of direct gel loading of PCR products.

Features of Platinum Taq Green Hot Start DNA Polymerase include:
• High specificity and increased yields with antibody-mediated hot-start PCR
• Convenient room temperature reaction setup
• Direct gel loading with green PCR buffer
• Versatile formulation for broad range of amplicons

About Platinum Taq DNA Polymerase
Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary antibodies that inhibit enzyme activity at room temperature, reducing nonspecific target amplification at lower temperatures. The polymerase is activated in a temperature-dependent manner (at 94°C) during the start of PCR. Once dissociated from the antibodies, the Taq DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5'→3' exonuclease activity (but not 3'→5' exonuclease activity).

Using Platinum Taq Green Hot Start DNA Polymerase
Platinum Taq Green Hot Start DNA Polymerase is provided with the optional KB Extender to get more versatility in PCR assays with long, GC-rich, or problematic amplicons. The green PCR buffer used with Platinum Taq DNA Polymerase allows for direct gel loading of PCR products. The dyes in the green buffer do not interfere with PCR performance and are compatible with downstream applications including DNA sequencing, ligation, and restriction digestion. Use Platinum Taq Green Hot Start DNA Polymerase for the amplification of DNA from complex genomic, viral, and plasmid templates, as well as in RT-PCR.

More Platinum DNA Polymerase products >
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Platinum™ Taq DNA Polymerase
Hot Start: Built-In Hot Start
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: Low
Reaction Format: Separate Components
Product Overhang: 3'-A
Reaction Speed: Standard
Final Product Size(s): 5 kb or less
Product Line: Platinum™
Product Size: 300 reactions
Shipping Condition: Dry Ice

Contents & storage

Each box contains:
• 60 µL Platinum Taq DNA Polymerase
• 2 x 1.25 mL 10X Green PCR Buffer, -Mg
• 1.25 mL 50 mM magnesium chloride
• 1.25 mL KB Extender

Store at -20°C in a non-frost-free freezer.

Documents

Manuals & protocols

Recommended products