Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Thermo Scientific Chemicals

Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic Jerrycan
Catalog number 119690050
Price (USD)
-
Quantity:
5 L
Packaging:
Plastic Jerrycan
Request bulk or custom format
Chemical Identifiers
CAS50-00-0
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer5 to 15 % methanol
Color scale=<10 APHA
Free acid (HCOOH)=<0.1 %
Appearance (Form)Clear solution
Titration35.0 to 39.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; that can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • It May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only