Formamidine acetate, 99%
Formamidine acetate, 99%
Formamidine acetate, 99%
Thermo Scientific Chemicals

Formamidine acetate, 99%

CAS: 3473-63-0 | CH4N2·C2H4O2 | 104.11 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 119711000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3473-63-0
IUPAC Nameacetic acid; methanimidamide
Molecular FormulaC3H8N2O2
InChI KeyXPOLVIIHTDKJRY-UHFFFAOYSA-N
SMILESNC=N.CC(O)=O
さらに表示
Appearance (Form)Crystalline powder or crystals
Titration with HClO4>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only