Formamidine acetate, 99%
Formamidine acetate, 99%
Formamidine acetate, 99%
Thermo Scientific Chemicals

Formamidine acetate, 99%

CAS: 3473-63-0 | CH4N2·C2H4O2 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 119715000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3473-63-0
IUPAC Nameacetic acid; methanimidamide
Molecular FormulaC3H8N2O2
InChI KeyXPOLVIIHTDKJRY-UHFFFAOYSA-N
SMILESNC=N.CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with HClO4>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only