Formamidinesulfinic acid, 99%
Formamidinesulfinic acid, 99%
Formamidinesulfinic acid, 99%
Thermo Scientific Chemicals

Formamidinesulfinic acid, 99%

CAS: 1758-73-2 | CH4N2O2S | 108.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
119725000500 gPlastic bottle
Catalog number 119725000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1758-73-2
IUPAC Namecarbamimidoylsulfinic acid
Molecular FormulaCH4N2O2S
InChI KeyFYOWZTWVYZOZSI-UHFFFAOYNA-N
SMILESNC(=N)S(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Water=<0.5 %
Appearance (Color)White to light yellow
Titration with NaOH>=98.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only