Furfuryl mercaptan, 97%
Furfuryl mercaptan, 97%
Furfuryl mercaptan, 97%
Thermo Scientific Chemicals

Furfuryl mercaptan, 97%

CAS: 98-02-2 | C5H6OS | 114.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 119811000
Price (EUR)
114,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
114,00
Each
Chemical Identifiers
CAS98-02-2
IUPAC Name(furan-2-yl)methanethiol
Molecular FormulaC5H6OS
InChI KeyZFFTZDQKIXPDAF-UHFFFAOYSA-N
SMILESSCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5290 to 1.5330 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only