Interested in a bulk or custom version of this product? Request pricing.

KEAP1 Recombinant Human Protein, N-His.GST.AVI Tag

Catalog number: 11981H29B5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Kelch-like ECH-Associated Protein 1 (KEAP1), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human KEAP1 (Q14145) (Gln 2-Cys 624) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus linked by the AVI tag.

Activity: This recombinant protein has not been tested.

Formulation: 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biologica Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: RP11-104M22.1, MGC126323, MGC126324
Gene ID: 9817
Gene Symbol: KEAP1
Molecular Weight: 99.2 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: Kelch-like ECH-Associated Protein 1
Protein Tag or Fusion: N-His, GST & AVI
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Research Category: Other Research
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols