Galvinoxyl free radical, 98%, Thermo Scientific Chemicals
Galvinoxyl free radical, 98%, Thermo Scientific Chemicals
Galvinoxyl free radical, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Galvinoxyl free radical, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
1198500505 gGlass bottle
Catalog number 119850050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2370-18-5
IUPAC Name2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC29H42O2
InChI KeyHRLDHROCDBYHAU-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point156°C to 160°C
Appearance (Color)Dark blue to purple
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Polymerization reagent
RUO – Research Use Only