Girard's Reagent T, 99%
Girard's Reagent T, 99%
Girard's Reagent T, 99%
Thermo Scientific Chemicals

Girard's Reagent T, 99%

CAS: 123-46-6 | C5H14ClN3O | 167.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 119872500
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS123-46-6
IUPAC Name[(hydrazinecarbonyl)methyl]trimethylazanium chloride
Molecular FormulaC5H14ClN3O
InChI KeyYSULOORXQBDPCU-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)(C)CC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Argentometric>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point187°C to 192°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only