D-Glucosamine hydrochloride, 98+%
D-Glucosamine hydrochloride, 98+%
D-Glucosamine hydrochloride, 98+%
Acros Organics

D-Glucosamine hydrochloride, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 119900025
also known as 11990
HSN 29329900
Price (INR)
136,261.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
136,261.00
Each
Chemical Identifiers
CAS66-84-2
IUPAC Namehydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
Molecular FormulaC6H14ClNO5
InChI KeyCBOJBBMQJBVCMW-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 % (On dry substance)
Arsenic (As)=<3 ppm
Specific optical rotation(after 3 h)
Specific optical rotation+70.0° to +73.0° (20°C, 589 nm) (c=2.5, H2O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only