SP Scienceware™ Conical Tube Holder
SP Scienceware™ Conical Tube Holder
SP Scienceware™ Conical Tube Holder
SP Scienceware™ Conical Tube Holder
Fisher Scientific™

SP Scienceware™ Conical Tube Holder

Allows user to clearly view the entire contents of tubes. Bel Art™ SP Scienceware™ Conical Tube Holder sets up in just seconds to hold either a 15ml or a 50ml conical tube.
Have Questions?
카탈로그 번호용도(장비)색상오토클레이브 가능물질
11999001
FISHERTOPSELLING-11999001-EACH으로도 사용됨
15 and 50mL Conical TubesTransparentAutoclavablePolypropylene
카탈로그 번호 11999001
FISHERTOPSELLING-11999001-EACH으로도 사용됨
제품 가격(KRW)
-
용도(장비):
15 and 50mL Conical Tubes
색상:
Transparent
오토클레이브 가능:
Autoclavable
물질:
Polypropylene
Allows user to clearly view the entire contents of tubes. Bel Art™ SP Scienceware™ Conical Tube Holder sets up in just seconds to hold either a 15ml or a 50ml conical tube.

  • Versatile holder supports either a 15 or 50mL conical tube while allowing a view of the entire tube contents
  • Move tube and rack with one hand by pinching the uprights
  • Insert or remove tube from the holder with one hand
  • Steam autoclavable at 121°C (250°F)
  • Chemical-resistant polypropylene

사양
제품 유형Test Tube Single Rack
오토클레이브 가능Autoclavable
색상Transparent
보유Tubes (15 or 50 mL)
물질Polypropylene
위치1
용도(장비)15 and 50mL Conical Tubes
Unit SizeEach