Glutaryl dichloride, 97%
Glutaryl dichloride, 97%
Glutaryl dichloride, 97%
Thermo Scientific Chemicals

Glutaryl dichloride, 97%

CAS: 2873-74-7 | C5H6Cl2O2 | 169.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 119991000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2873-74-7
IUPAC Namepentanedioyl dichloride
Molecular FormulaC5H6Cl2O2
InChI KeyYVOFTMXWTWHRBH-UHFFFAOYSA-N
SMILESClC(=O)CCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration RCOCl>=96.0 %
Refractive index1.4720 to 1.4760 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to red-brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only