Glycine methyl ester hydrochloride, 98%
Glycine methyl ester hydrochloride, 98%
Glycine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

Glycine methyl ester hydrochloride, 98%

CAS: 5680-79-5 | C3H8ClNO2 | 125.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 120105000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5680-79-5
IUPAC Namehydrogen methyl 2-aminoacetate chloride
Molecular FormulaC3H8ClNO2
InChI KeyCOQRGFWWJBEXRC-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC(=O)CN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Titration with HClO497.5 to 102.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only