Guanidine Carbonate, 99+%
Guanidine Carbonate, 99+%
Guanidine Carbonate, 99+%
Thermo Scientific Chemicals

Guanidine Carbonate, 99+%

CAS: 593-85-1 | 0·5 H2CO3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 120220010
Price (EUR)
76,80
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
76,80
Each
Chemical Identifiers
CAS593-85-1
IUPAC Namecarbonic acid; bis(guanidine)
Molecular FormulaC3H12N6O3
InChI KeySTIAPHVBRDNOAJ-UHFFFAOYSA-N
SMILESNC(N)=N.NC(N)=N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only