Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)
Thermo Scientific Chemicals

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry) | CAS: 50-01-1 | CH5N3·HCl | 95.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 120232500
Price (USD)
46.80
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
46.80
Each
Chemical Identifiers
CAS50-01-1
IUPAC Namehydrogen guanidine chloride
Molecular FormulaCH6ClN3
InChI KeyPJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Titration Argentometric=<0.5 % Ammonium chloride
Appearance (Form)Crystalline powder
Water=<0.3 %
Anticaking agent=<0.4 % (SiO2)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only