Nunc™ Cell Factory™ Racks and Carts
Actual product may vary
Thermo Scientific™
Nunc™ Cell Factory™ Racks and Carts
Hold up to four Cell Factory 40 or 12 Cell Facotry 10 systems with Nunc Cell Factory Racks and Carts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
120289Retrofit Clamping KitShaker
140503Side Loading Rack with CartCell Factory 40
140504Side Loading Rack with CartCell Factory 10
Catalog number 120289
Price (USD)
-
Product Type:
Retrofit Clamping Kit
For Use With (Equipment):
Shaker
Store and transport equipment effectively using Thermo Scientific™ Nunc™ Cell Factory™ Racks and Carts, which can hold up to four Cell Factory 40 or 12 Cell Factory 10 systems. These racks and carts are designed to integrate perfectly with all Nunc Cell Factory equipment.
  • Constructed of AISI 316 Stainless Steel
  • Designed for use in clean room environments
  • Side-loading design for easy loading and unloading
Specifications
DescriptionRetrofit Clamping Kit for Racks for use with Shaker
For Use With (Equipment)Shaker
MaterialStainless Steel (AISI 316)
Product TypeRetrofit Clamping Kit
Unit SizeEach