1-Hexadecanol, 96%
1-Hexadecanol, 96%
1-Hexadecanol, 96%
Thermo Scientific Chemicals

1-Hexadecanol, 96%

CAS: 36653-82-4 | C16H34O | 242.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Catalog number 120480010
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.60
Save 8.95
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.60
Save 8.95
Each
Chemical Identifiers
CAS36653-82-4
IUPAC Namehexadecan-1-ol
Molecular FormulaC16H34O
InChI KeyBXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or flakes or pellets
Appearance (Color)White to off-white
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only