1-Hexadecene, 92%, tech.
1-Hexadecene, 92%, tech.
1-Hexadecene, 92%, tech.
Thermo Scientific Chemicals

1-Hexadecene, 92%, tech.

CAS: 629-73-2 | C16H32 | 224.43 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1205000101 LGlass bottle
Catalog number 120500010
Price (EUR)
101,00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
101,00
Each
Chemical Identifiers
CAS629-73-2
IUPAC Namehexadec-1-ene
Molecular FormulaC16H32
InChI KeyGQEZCXVZFLOKMC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4390 to 1.4430 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=90 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only