2,4-Hexadienoic acid, 99%
2,4-Hexadienoic acid, 99%
2,4-Hexadienoic acid, 99%
Thermo Scientific Chemicals

2,4-Hexadienoic acid, 99%

CAS: 110-44-1 | C6H8O2 | 112.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 120532500
Price (EUR)
49,65
Online Exclusive
55,30
Save 5,65 (10%)
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
49,65
Online Exclusive
55,30
Save 5,65 (10%)
Each
Chemical Identifiers
CAS110-44-1
IUPAC Name(2E,4E)-hexa-2,4-dienoic acid
Molecular FormulaC6H8O2
InChI KeyWSWCOQWTEOXDQX-MQQKCMAXSA-N
SMILESC\C=C\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point132°C to 135°C
Titration with NaOH98.5 to 101.5 % (on anhydrous substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only