AIM V™ Medium, liquid (research grade), 500 mL - FAQs
AIM V™ Medium, liquid (research grade), 500 mL - FAQs
View additional product information for AIM V™ Medium, liquid (research grade) - FAQs (12055083, 12055091)
2 product FAQs found
What is the difference between AIM V Medium (Cat. Nos. 12055083, 12055091) and CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK)?
The difference is in the intended usage of the two media. AIM V Medium (Cat. Nos. 12055083, 12055091) is for research use only whereas CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK) is for research use or manufacturing of cell, gene, or tissue-based products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free medium would you recommend for culture and expansion of mouse T cells?
You can try using AIM V Medium (Cat. No. 12055083 or 12055091) for growth of mouse T cells. For mouse cells, we recommend adding 50 µM beta-mercaptoethanol (Cat. No. 21985023).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.