Lysis Buffer (R2 )

Catalog number: 12057022

Invitrogen™  Related applications: Automated Nucleic Acid Purification | DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Lysis Buffer (R2) is a component in the PureLink™ 96 Plasmid Purification System. The PureLink™ 96 Plasmid Purification System offers a unique approach to high throughput plasmid DNA isolation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Lysis Buffer
Product Size: 1 L
Shipping Condition: Room Temperature

Contents & storage

1 L volume The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.

Documents

Manuals & protocols