Hexamethylbenzene, 98+%
Hexamethylbenzene, 98+%
Hexamethylbenzene, 98+%
Hexamethylbenzene, 98+%
Thermo Scientific Chemicals

Hexamethylbenzene, 98+%

CAS: 87-85-4 | C12H18 | 162.27 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 120570250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS87-85-4
IUPAC Namehexamethylbenzene
Molecular FormulaC12H18
InChI KeyYUWFEBAXEOLKSG-UHFFFAOYSA-N
SMILESCC1=C(C)C(C)=C(C)C(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Form)Crystalline powder or crystals or needles
Melting point164°C to 168°C
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only