Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Thermo Scientific Chemicals

Hexamethylenetetramine, 99%

CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
檢視全部
Plastic Bottle
產品號碼 120610025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
化學識別
CAS100-97-0
IUPAC Name1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular FormulaC6H12N4
InChI KeyVKYKSIONXSXAKP-UHFFFAOYSA-N
SMILESC1N2CN3CN1CN(C2)C3
檢視更多
規格Specification Sheet規格表
Titration with HClO4>=98.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Ash=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only