1,6-Hexanediamine, 99.5+%
1,6-Hexanediamine, 99.5+%
1,6-Hexanediamine, 99.5+%
Acros Organics

1,6-Hexanediamine, 99.5+%

CAS: 124-09-4 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 120641000
HSN 29212200
Price (INR)
4,415.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,415.00
Each
Chemical Identifiers
CAS124-09-4
IUPAC Namehexane-1,6-diamine
Molecular FormulaC6H16N2
InChI KeyNAQMVNRVTILPCV-UHFFFAOYSA-N
SMILESNCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Semitransparent white to light yellow to beige
Appearance (Form)Crystalline low melting mass
Infrared spectrumConforms
Melting point38°C to 43°C
GC>=99.50 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only