Hexanoic acid, 99%
Hexanoic acid, 99%
Hexanoic acid, 99%
Thermo Scientific Chemicals

Hexanoic acid, 99%

CAS: 142-62-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 120702500
Price (USD)
51.90
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.90
Each
Chemical Identifiers
CAS142-62-1
IUPAC Namehexanoic acid
Molecular FormulaC6H12O2
InChI KeyFUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILESCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4150 to 1.4180 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only