Hexyl alcohol, 98%, pure
Hexyl alcohol, 98%, pure
Hexyl alcohol, 98%, pure
Thermo Scientific Chemicals

Hexyl alcohol, 98%, pure

CAS: 111-27-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 120790025
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Chemical Identifiers
CAS111-27-3
IUPAC Namehexan-1-ol
Molecular FormulaC6H14O
InChI KeyZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILESCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Color scale=<10 APHA
GC>=97.5 %
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
Acid value=<0.02 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only