L(+)-Histidine methyl ester dihydrochloride, 97%
L(+)-Histidine methyl ester dihydrochloride, 97%
L(+)-Histidine methyl ester dihydrochloride, 97%
Thermo Scientific Chemicals

L(+)-Histidine methyl ester dihydrochloride, 97%

CAS: 7389-87-9 | C7H11ClN3O2 | 204.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 120820050
Price (USD)
24.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.00
Save 4.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.00
Save 4.35
Each
Chemical Identifiers
CAS7389-87-9
IUPAC Namemethyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl
Molecular FormulaC7H11ClN3O2
InChI KeyVXXIPPPPKNWFLK-UHFFFAOYNA-N
SMILES[Cl].COC(=O)C(N)CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Color)White
Specific optical rotation+8° to +11° (20°C, 589 nm) (c=2, water)
Appearance (Form)Crystalline powder
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only