Interested in a bulk or custom version of this product? Request pricing.

BCL6 Recombinant Human Protein, N-Trx.His tag

Catalog number: 12083H30E25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

B-cell CLL/Lymphoma 6 (BCL6), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human BCL6 (P41182) N-terminal fragment (Met 1-Met 150) fused to a Trx and a polyhistidine tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 20 mM Tris, 10% glycerol, pH 8.0.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biologica Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Gene ID: 604
Gene Symbol: BCL6
Molecular Weight: 34.2 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Transcription Factors
Protein Form: Recombinant, Truncated
Protein Name: B-Cell Cll/Lymphoma 6
Protein Tag or Fusion: His Tag, Thioredoxin
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Oncology
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols